missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate monohydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog Number p-4407587
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
[Li+].[Li+].O.[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
[Li+].[Li+].O.[O-]S([O-])(=O)=O

Maximum level of impurities: Loss on drying at 150°C 13.0-15.0%, Insoluble matter 0.01%, Cl 0.002%, NO3 0.001%, Heavy Metals (as Pb) 0.001%, Fe 0.001%, K 0.05%, Na 0.05%

Lithium sulfate monohydrate is used in solar panels and potential for a new class of battery. It is also used to treat bipolar disorder and in ultrasound-type non-destructive testing. It is also used as an additive for portland cement to speed up the curing. It is also involved as a catalyst in the elimination reaction for the conversion of n-butyl bromide to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130°C-H2O
Color Colorless
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Soluble in water. Insoluble in ethanol,acetone and pyridine.
IUPAC Name dilithium;sulfate;hydrate
Formula Weight 127.96 (109.94 Anhydrous)
Grade ACS
Odor Odorless
Appearance Colorless granules
Density 2.06
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'