Learn More
Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol, Thermo Scientific™
Catalog Number p-7051975
Click to view available options
Quantity:
25 mL
100 mL
127503-04-2
MFCD00210630
lithium tantalum ethoxide
127503-04-2
MFCD00210630
lithium tantalum ethoxide
Precursor for Lithium tantalum oxide films
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Color Yellow
Odor Alcohol-like
Linear Formula LiTa(OCH2CH3)6
Quantity 25 mL
UN Number UN1170
Sensitivity Moisture Sensitive
Formula Weight 458.04
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lithium tantalum ethoxide
Show More Show Less
compliance-icons
Product Identifier
  • Lithium tantalum ethoxide 5% w/v in ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Lithium tantalum ethoxide