missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4813518
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy
1907-33-1, 96-47-9
C4H9LiO
80.05
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
This item is not returnable. View return policy
1907-33-1, 96-47-9
C4H9LiO
80.05
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Linear Formula LiOCH(CH3)3
Flash Point -11°C
Formula Weight 80.05
Specific Gravity 0.86
Density 0.8600g/mL
Chemical Name or Material Lithium tert-butoxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium tert-butoxide, 8% in 2-MeTHF
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'