missing translation for 'onlineSavingsMsg'
Learn More

Lithium wire, 3.2mm (0.125in) dia, 99.8%, Na 0.03% max, Thermo Scientific Chemicals

Catalog Number p-6827149
Click to view available options
Quantity:
25 g
4 x 25 g
7439-93-2
Li
6.94
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
3028194
CHEBI:30145
lithium
[Li]
7439-93-2
Li
6.94
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
3028194
CHEBI:30145
lithium
[Li]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.53
Assay Percent Range 99.8%
Quantity 25 g
Formula Weight 6.941
Form Annealed Wire
Chemical Name or Material Lithium, 3.2mm (0.125 in.) Dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'