Learn More
Thermo Scientific™ LN2 Level Monitor
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Locator 4 Plus
Locator 8 Plus
Locator Jr. Plus
Voltage:
100/240 V
230 V
Unit Size:
Each
Description
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
- Minimizes conduction and liquid nitrogen evaporation
- Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
- Audible and visual alarms signal low level liquid nitrogen conditions
Specifications
Specifications
| Description | Level Monitor |
| Type | LN2 Measuring Device |
| For Use With (Equipment) | Locator 8 Plus |
| Monitoring Options | Alarm |
| Voltage | 100/240 V |