Description
Add an extra level of protection to the laboratory in the event of excessive liquid nitrogen loss during dewar storage with Thermo Scientific™ LN2 Low-Level Alarms, for use with any standard-sized LN2 container.
Low level alarm (120V) with sensor for any standard-sized LN2 container.
Includes:
Dry contacts for remote alarm
Specifications
Specifications
| Description | Low-level alarm with dry contacts for connection to telephone dialing system or remote alarm, customer installed |
| Type | LN2 Low-Level Alarm |
| For Use With (Equipment) | Any Standard-Sized LN2 Container |
| Monitoring Options | Alarm |
| Voltage | 120 V |