Learn More
Astell Scientific™ Load Support Plate
Support shelf to expand autoclave capacity. Astell Scientific™ Load Support Plate accommodates additional Duran bottles without the need for baskets.
Brand:  Astell Scientific™ AAN002
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 12708915
  • 201.00€ / Each
Estimated Shipment
12-08-2021
Add to basket
Description
Description
  • Expands autoclave loading capacity.
  • Supports additional Duran bottles without the need for baskets.
  • Stainless steel construction.
Specifications
Specifications
AMB420 and AMB220 autoclaves
Load support plate
Stainless steel
1Each