Learn More
Loctite™ 7100 Leak Detector
Product Code. 11930207
Encompass
Click to view available options
Volume (Metric):
400 mL
Unit Size:
400mL
Product Code. 11930207
Brand: Loctite 250000
Specifications
Product Type Leak Detector
Format Liquid
For Use With (Application) Suitable for all gases, even oxygen
Boiling Point 100°C
Color Colorless
Pressure 4000mbar
Volume (Metric) 400 mL
compliance-icons
Product Identifier
  • LOCTITE SF 7100
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H229-Pressurised container: May burst if heated.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P251-Do not pierce or burn, even after use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.