Learn More
Agilent BioTek LogPhase 600 Product Qualification Package
Product Code. 17132089 Shop All Agilent BioTek Products
Change view
Click to view available options
:
LogPhase 600 Product Qualification Package
Product Code.
17132089 LogPhase 600 Product Qualification Package
1 options
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Product Code. 17132089
Brand: Agilent BioTek 1800547
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
LogPhase 600 Product Qualification Package is for use with LogPhase 600 Microbiology Reader
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LogPhase 600 Microbiology Reader; Requires Gen5 309 or Higher; Agilent BioTek Microplate Washer and Reader Instruments
Product Type Product Qualification Package