Learn More
Lonza™ SYPRO™ Ruby Protein Blot Stain
Description
Does not interfere with subsequent immunodetection.
Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.
Specifications
Specifications
| Color | Ruby |
| Form | Liquid |
| For Use With (Application) | Fluorogenic, chemiluminescent and colorimetric detection techniques |
| Quantity | 200 mL |
| Product Type | Protein Stain |