Learn More
Copan Italia™ Loop in Resealable Pouch
Product Code. 11706548
Click to view available options
Volume:
1 μL
Unit Size:
Pack of 4000
Product Code. 11706548
Brand: Copan Italia™ 179CSR40
Only null left
Add to basket
Add to basket
Use Copan Italia™ Loop in Resealable Pouch as a flexible inoculator for brittle agar. The light blue loop is disposable, calibrated and flexible.
  • DIsposable
  • Calibrated
  • 4000Pack
  • Light Blue
Specifications
Disposable Single-use
Volume 1 μL
Material Flexible
Sterility Gamma-irradiated
Color Light Blue
Product Type Inoculating Loop
Quantity 4000/Pk.
Calibrated Yes