missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loratadine Epoxide N-Oxide, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Loratadine Epoxide N-Oxide
Specifications
Specifications
| Chemical Name or Material | Loratadine Epoxide N-Oxide |
| CAS | 1432057-93-6 |
| Analyte or Component Names | Loratadine Epoxide N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H23 Cl N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Loratadine Epoxide N-Oxide |
| InChI Formula | InChI=1S/C22H23ClN2O4/c1-2-28-20(26)24-12-9-21(10-13-24)22(29-21)18-8-7-17(23)14-16(18)6-5-15-4-3-11-25(27)19(15)22/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3 |
| SMILES | CCOC(=O)N1CCC2(CC1)OC23c4ccc(Cl)cc4CCc5ccc[n+]([O-])c35 |
| Molecular Weight (g/mol) | 414.88 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction