missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Aqua Comparitor Color Discs: pH
For testing water with the Aqua Comparator
26.10€
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
11715448
|
Lovibond™
146100 |
pH Color Disc | 6.5 to 8.4 pH |
26.10€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
11725448
|
Lovibond™
146110 |
pH Bromcresol Purple Color Disc | 5.2 to 6.8 pH |
26.10€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
11735448
|
Lovibond™
146120 |
pH Bromthymol Blue Color Disc | 6 to 7.6 pH |
26.10€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
11745448
|
Lovibond™
146130 |
Universal pH Color Disc | 4 to 10 pH |
26.10€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |