Learn More
Color Standard: Platinum-Cobalt Color 100, Lovibond™
Brand: Lovibond Tintometer 134190
Additional Details : Weight : 0.50000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Specifications
Color Reference Standard |
Safety and Handling
- reference standard Pt-Co 100/platinum cobalt 100
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : hydrochlroic acid, cobalt dichloride hexahydrochlorate