Learn More
Lovibond™ Color of Water Comparator Disc for 700 Nesslerizer Comparator
Brand:  Lovibond™ 284160
Additional Details : Weight : 0.80000kg
Product Code. 11934744
  • 278.00€ / Each
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
Color of Water Comparator Disc for 700 Nesslerizer Comparator
Specifications
Specifications
Measuring the color of potable water in terms of an easily reproducible color standard
30 to 70 Color Units
Color of Water Comparator Disc
700 Nesslerizer comparator
Pt-Co or APHA