Learn More
Lovibond™ Color of Water Comparator Disc for 711 Nesslerizer Comparator
Brand:  Lovibond™ 284150
Additional Details : Weight : 0.99000kg
Product Code. 10579583
  • 278.00€ / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
Color of Water Comparator Disc for 711 Nesslerizer Comparator
Specifications
Specifications
Measuring the color of potable water in terms of an easily reproducible color standard
0 to 30 Color Units
Color of Water Comparator Disc
711 Nesslerizer comparator
Pt-Co or APHA