Learn More
Lovibond™ Color of Water Comparator Disc for 711 Nesslerizer Comparator
Brand: Lovibond™ 284150
Additional Details : Weight : 0.99000kg
Description
Color of Water Comparator Disc for 711 Nesslerizer Comparator
Specifications
Measuring the color of potable water in terms of an easily reproducible color standard | |
0 to 30 Color Units | |
Color of Water Comparator Disc |
711 Nesslerizer comparator | |
Pt-Co or APHA |