missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Colorimeter Fluoride Solution Reagent: SPADNS

For use with Fluoride Aqua Comparator Test Kit, MD600, MD100 Fluoride colorimeters, MD600 Multi-Parameter Colorimeters, Multidirect and Spectrodirect Spectrophotometer, XD7000

Brand:  Lovibond™ 467482

Additional Details : Weight : 0.30000kg Transport : UN number : 1789 Chem class : 8 Pack group : III

Product Code. 15352045

  • 128.00€ / 500mL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For use with Fluoride Aqua Comparator Test Kit, SC450, MD100 Fluoride colorimeters, MC500, MD600 Multi-Parameter Colorimeters and SP600 Spectrophotometer.
Specifications

Specifications

SPADNS Colorimeter Reagent
Fluoride Aqua Comparator Test Kit, SC450, MD100 Fluoride colorimeters, MC500, MD600 Multi-Parameter Colorimeters and SP600 Spectrophotometer.
500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SPADNS Reagent

Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid

SDS
Documents

Documents

Certificates
Promotions

Promotions