missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Phosphonate Reagent Set

Product Code. 11759059
Click to view available options
Range:
0 to 2.5mg/L
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11759059

Brand: Lovibond™ undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used with MC500 Colorimeter and SP600 Spectrophotometer

  • Pre-measured
  • Easy Open notch
  • Beveled edge for easy dispense and pour

Specifications

For Use With (Application) Aqua comparator™ test kit
For Use With (Equipment) SC450 phosphate and MC500 colorimeters, SP600 spectrophotometer
Includes 10mL sample
Range 0 to 2.5mg/L
Type Phosphate 3 Reagent
compliance-icons compliance-icons
Product Identifier
  • X1000 REACTIF VARIO PHOS3 10ML
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : dipotassium disulphate, potassium antimony(III) oxide tartrate trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.