missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Reagent Set: Total and Acid Hydrolyzable Phosphorus
Click to view available options
Range:
0.02 to 1.1 mg/L P, 0.06 to 5 mg/L PO4
Unit Size:
Pack of 50
Description
Includes
Acid Reagent Vial (50ct), Phosphate 3 Reagent (10mL sample, 50 pack), Potassium Persulfate Reagent (10mL sample, 50 pack), Sodium hydroxide (1.54 N, 100mL), Deionized water (100mL)
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Orbeco-Hellige and Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor |
| Includes | 50 ct. acid reagent vial, 10mL phosphate 3 reagent (50 pack), 10mL potassium persulfate reagent (50 pack), 1.54N 100mL sodium hydroxide, 1N 100mL sodium hydroxide, 100mL deionized water |
| No. of Tests | 50 |
| Range | 0.02 to 1.1 mg/L P, 0.06 to 5 mg/L PO4 |
| Test Method | Acid digestion/Ascorbic Acid |
| Type | Total Phosphate and Acid Hydrolysable Reagent Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction