Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Click to view available options
Quantity:
100 Pack
100 mL
15 mL
200 Pack
250 Pack
Type:
DEHA Test Solution
DPD Glycine Test Tablets
DPD Glycine Test Tablets (for Cl separation)
EDTA Test Tablets
pH-Universal
Format:
Liquid
Tablet
Unit Size:
100 tablets
100mL
15mL
Each
Pack of 100
Pack of 250
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Specifications
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | Colorimeter and spectrophotometer |
| Quantity | 200 Pack |
| Type | DPD Glycine Test Tablets |