Learn More
Porcelaines Avignon™ Low Form Porcelain Crucibles
Product Code. p-8000832
Click to view available options
Capacity (Metric):
3 mL
6 mL
14 mL
25 mL
40 mL
80 mL
130 mL
250 mL
Unit Size:
Pack of 6
Product Code. 11748473
Brand: Porcelaines Avignon™ 11306
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Heat and cool substances in the laboratory using Porcelaines Avignon™ Low Form Porcelain Crucibles. They are available in various capacities.
  • Low walls
  • Withstand high temperatures and rapid temperature changes
  • Porcelain
Specifications
Material Porcelain
Finish Glazed
Capacity (Metric) 40 mL
Diameter (Metric) Outer 55mm
Height (Metric) 36mm
Product Type Crucible