Learn More
Thermo Scientific Chemicals Low Form Zirconium Crucible
Product Code. 15493615
Click to view available options
Capacity (Metric):
15 mL
20 mL
25 mL
35 mL
55 mL
75 mL
100 mL
250 mL
500 mL
Diameter (Metric) Outer:
33mm
45mm
46mm
47mm
51mm
59mm
82mm
102mm
Height (Metric):
23mm
30mm
35mm
43mm
46mm
60mm
66mm
Unit Size:
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15493615
Brand: Thermo Scientific Chemicals 036018.KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Low Form
Material Zirconium
Capacity (Metric) 75 mL
Diameter (Metric) Outer 51mm
Height (Metric) 43mm
Product Type Zirconium Crucible
Diameter (Metric) Base 41mm