missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Low Ionic Strength Buffer Solution pH 6.96 at 20 °C, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Tested by ISO 17025 accredited test method
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| pH | 6.96 ±0.04 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | HDPE bottle GL45 |
| Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction