Learn More
Techne™ Temperature Bath Fluid
Enhance the performance of your Techne baths and circulators
Brand: Techne™ FHTFLO
Additional Details : Weight : 0.10000kg
Description
- Enhances bath temperature uniformity and stability
- Safe and non-toxic high temperature bath fluid that is versatile and cost effective
Specifications
≤118°C | |
177°C | |
18 L |
Polymer and Gasket | |
Low Temperature Heat Transfer Fluid |