Learn More
Techne™ Temperature Bath Fluid
Enhance the performance of your Techne baths and circulators
Brand:  Techne™ FHTFLO
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15117046
  • 674.00€ / Each
Estimated Shipment
04-06-2021
Add to basket
Description
Description
  • Enhances bath temperature uniformity and stability
  • Safe and non-toxic high temperature bath fluid that is versatile and cost effective
Specifications
Specifications
≤118°C
177°C
18 L
Polymer and Gasket
Low Temperature Heat Transfer Fluid