Learn More
Fisherbrand™ Teclu Burner with NF Connection, 13mm Opening
Shop All Fisherbrand Products
Click to view available options
Gas Type:
Butane, LPG, Propane
Methane, Natural Gas
Unit Size:
Each
Specifications
Specifications
| Product Type | Teclu Burner |
| For Use With (Equipment) | NF Screwing G 0.5 in Connector |
| Gas Type | Methane, Natural Gas |
| Diameter (Metric) | 13 mm |
| For Use With (Application) | Glasswork and soldering |
| Height (Metric) | 150 mm |
| Includes | NF Connector |
| No. per Pack | 1 Ea. |