Learn More
Corning™ LSE™ Shaking Incubator Accessory, Plastic Clamps
Product Code. 16279601 Shop All Corning Products
Click to view available options
Holds:
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 16279601
Brand: Corning™ 480150
Only null left
Add to basket
Add to basket
For use with LSE Shaking Incubator
Constructed of polypropylene.
TRUSTED_SUSTAINABILITY
Specifications
Holds 250 mL
Product Type Clamp
For Use With (Equipment) LSE Benchtop Shaking Incubator