Learn More
Poulten & Graf GmbH Luer Lock Glass and Metal Syringe
Click to view available options
Volume (Metric):
10 mL
2 mL
30 mL
500 to 5000 μL
Unit Size:
Each
Pack of 50
Description
Made of metal and glass
Amber graduations
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Syringe |
| Quantity | 1/Ea. |
| Volume (Metric) | 2 mL |