Learn More
Emma Safety Footwear Lukas XXD6T Safety Shoes
Click to view available options
Size:
39
47
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- Steel toe cap
- PU outer toe cap
- Extremely resilient reinforced sole
- Hydro-Tec™ Sanitized Silver
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Lukas XXD6T |
| Liner Material | Hydro-Tec™ Sanitized Silver |
| Material | Leather |
| Size | 39 |
| Toe Material | Steel |