missing translation for 'onlineSavingsMsg'
Learn More

Luminol, TRC

Product Code. 30469193
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
Product Code. Quantity unitSize
30469193 10 g 10g
30475599 1 g 1g
30481435 50 g 50g
3 options
This item is not returnable. View return policy

Product Code. 30469193

Brand: Toronto Research Chemicals TRCL47445010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-31-3
Formula Weight 177.0538
InChI Formula InChI=1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
Molecular Formula C8 H7 N3 O2
Molecular Weight (g/mol) 177.16
SMILES Nc1cccc2C(=O)NNC(=O)c12
Synonym 1,4-Phthalazinedione, 5-amino-2,3-dihydro-, 5-Amino-2,3-dihydro-1,4-phthalazinedione, 3-Aminophthalhydrazide, 3-Aminophthalic acid hydrazide, 3-Aminophthalic hydrazide, 5-Amino-1,4-dihydroxyphthalazine, Diogenes reagent, Luminol, NSC 5064
Chemical Name or Material Luminol

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.