missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 2603618
GREENER_CHOICE

Product Code. p-7161650
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797976

Brand: Tocris Bioscience™ 6454/10

This item is not returnable. View return policy

Potent and selective Chk1 inhibitor (IC50 = 7 nM)

LY 2603618 is a potent and selective Chk1 inhibitor (IC50 = 7 nM). Inhibits Chk1 autophosphorylation. Induces cell cycle arrest in the G2/M phase and induces the DNA damage response in lung cancer cells. Inhibits growth of NSCLC xenografts in mice when administered in combination with Pemetrexed (Cat.No. 6185).

Specifications

Chemical Name or Material N-[5-Bromo-4-methyl-2-[(2S)-2-morpholinylmethoxy)phenyl]-N'-(5-methyl-2-pyrazinyl)urea
CAS 911222-45-2
Quantity 10 mg
Molecular Formula C18H22BrN5O3
Formula Weight 436.3
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.