missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 2886721 Hydrochloride
GREENER_CHOICE

Product Code. 18769916
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18769916

Brand: Tocris Bioscience™ 7081/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective β-secretase (BACE) inhibitor

LY 2886721 Hydrochloride is a potent and selective β-secretase (BACE) inhibitor (IC50 values are 10.2 and 20.3 nM for human BACE2 and BACE1, respectively). Displays >5,000-fold selectivity for BACE over other proteases including cathepsin D, pepsin and renin. Inhibits Aβ1-40 and Aβ1-42 production in cells expressing mutated APP. Reduces hippocampal and cortical Aβ and sAPPβ levels in an Alzheimer's disease mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[(4 AS,7 AS)-2-Amino-4 A,5-dihydro-4 H-furo[3,4-d][1,3 ]thiazin-7 A(7 H)-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide hydrochloride
CAS 1262036-49-6
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C18H16F2N4O2S.HCl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.