missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Functional Grade, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

75.50€ - 2700.00€

Specifications

Antigen Ly-6G/Ly-6C
Clone RB6-8C5
Concentration 1 mg/mL
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15258697
View Documents Promotion Details
Affymetrix eBioscience
16-5931-82
100 μg
166.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15268697
View Documents Promotion Details
Affymetrix eBioscience
16-5931-85
500 μg
282.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15278697
View Documents Promotion Details
Affymetrix eBioscience
16-5931-86
1 mg
518.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15537796
View Documents Promotion Details
Affymetrix eBioscience
16-5931-95
10 mg
2700.00€
10mg
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15248697
View Documents Promotion Details
Affymetrix eBioscience
16-5931-81
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets. Applications Reported: The RB6-8C5 antibody has been reported for use in flow cytometric analysis. It has also been reported in cell depletion. Applications Tested: The RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Endotoxin: Less than 0.

Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications

Specifications

Ly-6G/Ly-6C
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN
Ly6c1, Ly6c2, Ly6g
Primary
4° C
RB6-8C5
Flow Cytometry, Functional Assay
Functional Grade
Rat
Mouse
P0CW02, P0CW03, P35461
100041546, 17067, 546644
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.