Learn More
Invitrogen™ Lysing Solution, Whole Blood Lysing Solution
Description
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Quantity | 25 mL |
| Volume (Metric) | 25 mL |
| Reagent Type | Whole Blood Lysis Buffer |
| Shipping Condition | Room Temperature |
| For Use With (Application) | Flow Cytometry |
| Product Type | Whole Blood Lysing Solution |