missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lysis Buffer, FS is designed specifically to lyse microbial organisms from food and environmental samples and prepare the microbial DNA for subsequent downstream analysis by real-time PCR.
This lysis buffer can be used in conjunction with Proteinase K, FS in the Pathatrix™ Auto Instrument or RapidFinder™ Direct Lysis workflows.
When used as a part of the Pathatrix™ Auto Instrument workflow, one bottle of Lysis Buffer, FS is sufficient for 700 reactions.
When used as a part of the RapidFinder™ Direct Lysis workflow, one bottle of Lysis Buffer, FS is sufficient for 100 reactions.
This lysis buffer can be used in conjunction with Proteinase K, FS in the Pathatrix™ Auto Instrument or RapidFinder™ Direct Lysis workflows.
When used as a part of the Pathatrix™ Auto Instrument workflow, one bottle of Lysis Buffer, FS is sufficient for 700 reactions.
When used as a part of the RapidFinder™ Direct Lysis workflow, one bottle of Lysis Buffer, FS is sufficient for 100 reactions.
Specifications
Specifications
| Density | 0.99 to 1.01g/mL |
| Description | Lysis Buffer, FS, 7mL, Liquid, Store at 4°C, 0.99 to 1.01g/mL Density, 1.330 to 1.340 Refractive Index, Water Soluble |
| For Use With (Application) | Clinical |
| Physical Form | Liquid |
| Quantity | 7 mL |
| Solubility Information | Water soluble |
For Testing of Food and Environmental Samples Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?