missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysis Solution for GeneJET Genomic DNA Purification Kit, Thermo Scientific™
Shop All Thermo Scientific ProductsDescription
Thermo Scientific GeneJET Lysis Solution is a component of the GeneJET Genomic DNA Purification Kit (K0721/K0722) and may be purchased separately.
Specifications
Specifications
| Chemical Name or Material | Lysis buffer |
| Description | Lysis Solution For GeneJET Genomic DNA Purification Kit, 60mL, Liquid, Lysis Buffer, Used For DNA Extraction |
| For Use With (Application) | DNA and RNA purification and analysis, DNA extraction, genomic DNA purification, gDNA from animal tissues and cells, gDNA from bacteria, gDNA from buccal cells, gDNA from blood (medium-scale, 100μL-1mL) |
| Physical Form | Liquid |
| Quantity | 60 mL |
| Formulation | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?