missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol Purple sodium salt, 0.04% w/v aq. soln., Thermo Scientific™
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Concentration | 0.04% w/v aq. soln. |
Formula Weight | 404.42 |
Quantity | 100 mL |
Physical Form | Liquid |
Chemical Name or Material | m-Cresol Purple sodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'