missing translation for 'onlineSavingsMsg'
Learn More

M-PEG4-thiol, TRC

Product Code. 30412811
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
Product Code. Quantity unitSize
30412811 200 mg 200mg
1 options
This item is not returnable. View return policy

Product Code. 30412811

Brand: Toronto Research Chemicals TRCP999268200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material M-PEG4-thiol
CAS 52190-55-3
Molecular Formula C9H20O4S
Quantity 200 mg
Synonym 2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethanethiol, 3,6,9,12-Tetraoxatridecane-1-thiol
InChI Formula InChI=1S/C9H20O4S/c1-10-2-3-11-4-5-12-6-7-13-8-9-14/h14H,2-9H2,1H3
SMILES COCCOCCOCCOCCS
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanethiol
Molecular Weight (g/mol) 224.32
Formula Weight 224.1082
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.