Learn More
Brady™ M410 Label Printer EU and BWS Suite
Product Code. 18744604
Click to view available options
Color:
Yellow, Black
Unit Size:
1 set
Regulatory: This product is under eco-contribution tax.
Product Code. 18744604
Brand: Brady™ 322717
Regulatory: This product is under eco-contribution tax.
Specifically engineered for premium 25.40 mm printing, the M410 Label Printer is an everyday workhorse for DataCom, electrical, and lab professionals.
  • Print pre-sized or continuous labels up to 25.40 mm wide with edge-to-edge printing
  • 110+ hassle-free drop-in cartridges with 12+ purpose-built materials to take on whatever your work demands
  • Get seamless cartridge changes with an easy-swap locking mechanism while a window in the door allows you to easily remember what's installed
  • Create and print two ways - the intuitive QWERTY keypad or a PC
  • Designed to withstand 1.2 m drops and challenging work environments
  • Enhanced exterior coloring for jobsite visibility
  • Ships with a licence for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
  • Backlit LCD graphics display for easier labelling in low-lit areas
Specifications
Product Type Printer
Color Yellow, Black
Display Type None
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 3.50 in.
Width (English) 6 in.
Length (Metric) 88.90 mm
Width (Metric) 152.40 mm
Height (English) 11.50 in.
Height (Metric) 292.10 mm
Color Capability Monocolor
Description M410 Label Printer EU and Brady Workstation Suite
Includes M410 Printer, Rechargeable NiMH Battery Pack, an AC Adapter, B-595 Vinyl label Cartridge, a Quick start Guide, a USB Cable, a Brady WorkstationBasic Design Software Suite and a USB Flash Drive with Driver and User Manual.
Languages Bulgarian, Chinese (Traditional and Simplified), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Italian, Japanese, Norwegian (Bokmal), Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Swedish, Turkish, Hungarian
Memory 48 MB
Model M410
Print Speed 33.02 mm/s
Resolution 300 dpi
Connector Types USB
Weight (English) 2.6 lb.
Weight (Metric) 1.18 kg
Certifications/Compliance CE, UKCA
For Use With (Application) Asset Tracking, Component and Equipment Labelling, Data and Telecommunications Labelling, Electrical Labelling, Facility Identification, General Identification, Inventory and Inspection Labelling, Laboratory Labelling, Wire and Cable Labelling
Quantity 1 Ea.
Warranty 2 Years
Power Supply Details AC Adapter, Rechargeable Battery
Voltage 100/240 V
Alternate Part Number M410-EU-BWS