missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar (Dehydrated)

Product Code. p-10710000
Click to view available options
Quantity:
500 g
2.5kg
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 10661675

Brand: Thermo Scientific™ CM0007B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this chromogenic medium.

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with Thermo Scientific™ Oxoid™ MacConkey Agar (Dehydrated). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water.

Use MacConkey Agar for the cultivation of pathogens which may be present in a variety of specimens.

  • Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphylococcus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
  • Recommended by World Health Organizaton1, the Department of Health2.

Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens such as urine, feces and wound swabs. While it is selective it does not suppress a mixed bacterial flora to the same extent as other inhibitory media (including other MacConkey agars). MacConkey Agar should be used in parallel with other selective indicator media such as Desoxycholate Citrate Agar, Bismuth Sulphite Agar, Brilliant Green Agar and Brilliant Green Bile (2%) Broth, and a non-selective medium such as Blood Agar.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description MacConkey Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.