missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MAD2L1 (Human) Recombinant Protein

Human MAD2L1 full-length ORF ( AAH00356, 1 a.a. - 205 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004085-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157511

  • 528.00€ / 25µg
Estimated Shipment: 09-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MALQLSREQGITLRGSAEIVAEFFSFGINSILYQRGIYPSETFTRVQKYGLTLLVTTDLELIKYLNNVVEQLKDWLYKCSVQKLVVVISNIESGEVLERWQFDIECDKTAKDDSAPREKSQKAIQDEIRSVIRQITATVTFLPLLEVSCSFDLLIYTDKDLVVPEKWEESGPQFITNSEEVRLRSFTTTIHKVNSMVAYKIPVND
Specifications

Specifications

AAH00356
MAD2 mitotic arrest deficient-like 1 (yeast)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAD2L1
GST
4085
Wheat germ expression system
25 μg
HSMAD2, MAD2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions