Learn More
R&D Systems™ MagCellect Streptavidin Ferrofluid
Click to view available options
Quantity:
1 Kit
Unit Size:
1mL
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| For Use With (Application) | Cell Isolation |
| Product Type | Streptavidin Ferrofluid |
| Quantity | 1 Kit |