missing translation for 'onlineSavingsMsg'
Learn More

Magnesium, 99.9+%, (trace metal basis), turnings, Thermo Scientific Chemicals

Catalog Number p-222356
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
7439-95-4
Mg
24.30
MFCD00085308
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]
7439-95-4
Mg
24.30
MFCD00085308
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
5462224
CHEBI:25107
magnesium
[Mg]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 651.0°C
Boiling Point 1107.0°C
Density 1.74g/mL
Assay Percent Range 99.9+%
Linear Formula Mg
Quantity 500 g
Fieser 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
Flash Point 500°C
Formula Weight 24.3
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Form Turnings
Chemical Name or Material Magnesium, 99.9+%
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
  • Self-heating substance Category 2
Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'