missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium chloride hexahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | MgCl2 · 6H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 203.3g/mol |
Percent Purity | 99 to 102% |
Chemical Name or Material | Magnesium chloride hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'