missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, purum p.a, ≥98.0% (KT), Honeywell™

Product Code. 15676060 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 15676060

Brand: Honeywell Research Chemicals 630651KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color Colorless
pH 5.0 to 6.0 (at 20°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity ≥98.0% (KT)
Packaging Poly bottle
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.