missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, Honeywell Fluka™

Catalog Number 15652180
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
10034-81-8
Cl2MgO8
223.197
MFCD00011102
MPCRDALPQLDDFX-UHFFFAOYSA-L
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
24840
magnesium;diperchlorate
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
This item is not returnable. View return policy
10034-81-8
Cl2MgO8
223.197
MFCD00011102
MPCRDALPQLDDFX-UHFFFAOYSA-L
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
24840
magnesium;diperchlorate
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 5 g
Linear Formula Mg(ClO4)2
UN Number 1475
Formula Weight 223.21g/mol
Grade ACS Reagent
Chemical Name or Material Magnesium Perchlorate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'