missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate Heptahydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
2.5kg
Specifications
Specifications
| Melting Point | 770°C |
| Color | White |
| pH | 5 to 8.2 |
| Physical Form | Solid |
| Quantity | 1 kg |
| Formula Weight | 246.48g/mol |
| Packaging | Plastic powder jar |
| Vapor Pressure | <0.1mmHg at 20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction