missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Magnetic Plastic Stickers for the Transport of Infectious Substances
Brand: Brady™ 258970
Additional Details : Weight : 0.10000kg
Specifications
Magnetic Plastic | |
Permanent Acrylic | |
297.00 mm | |
6 |
1 Ea. | |
297 mm | |
Rhomboid | |
Black on White |