missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Magnetic Stirrer TM 96

Product Code. 10739371 Shop All SI Analytics Products
Click to view available options
Model:
TM 96 TZ
Unit Size:
Each
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

Product Code. 10739371

Brand: SI Analytics™ 285223253

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

SI Analytics Magnetic Stirrer TM 96 for glass vessels up to maximal 500 mL.

  • Magnetic stirrer TM 96 for glass vessels up to maximum of 500 mL
  • Speed can be adjusted without discrete steps
  • Connection to TITRONIC™ universal, TITRONIC™ 97, TITRONIC™ 96, TITRONIC™ basic, TitroLine™ 96, TitroLine™ easy
TRUSTED_SUSTAINABILITY

Specifications

Model TM 96 TZ
Type Magnetic Stirrer
Weight (Metric) 0.80 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.