missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
Description
Includes
- MagniSort Anti-Mouse CD335 (NKp46) Biotin : 200 tests, 20μL/test
- MagniSort Positive Selection Beads A
- CD335+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD335+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (Integrin alpha 2), clone DX5, or Anti-Mouse NK1.1, clone PK136, in C57BL, FVB/N and NZB mice
Specifications
Specifications
| Content And Storage | 2° to 8°C. Do not freeze. |
| For Use With (Application) | Magnetic Cell Separation |
| Format | Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer |
| Quantity | 200 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction