Learn More
Invitrogen™ MagniSort™ Mouse Hematopoietic Lineage Depletion Kit
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Description
Includes
- MagniSort Mouse Hematopoietic Lineage Antibody Cocktail : 200 tests, 20μL/test
- MagniSort Negative Selection Beads B
- Lineage committed cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the lineage negative cells can be separated from lineage committed cells by decanting
- After depletion, the enrichment of multi-potent progenitor cells can be verified by staining with Anti-Mouse CD34 (clone RAM34), and the enrichment of CD34- hematopoietic stem cell can be verified by additional staining with CD150 (clone mShad150), CD48 (clone HM48-1)
Anti-Mouse CD2 Biotin
Anti-Mouse CD3 Biotin
Anti-Mouse CD5 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD45R Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
Specifications
Specifications
| Cell Type | Hematopoietic Cells |
| Product Type | Cell Depletion Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Mouse |